2-Amino-m-cresol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

39841-0050 39841-0250
TS39841-0050EA 79 USD
TS39841-0050 TS39841-0250
2-Amino-m-cresol 96%
2-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00075082
CAS Number: 2835-97-4

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 149 to 152 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR