4-tert-Butyltoluene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-Butyl)-4-methylbenzene

29632-0010 29632-0025 29632-0250
TS29632-0010EA 116 USD
TS29632-0010 TS29632-0025 TS29632-0250
4-tert-Butyltoluene 96%
4-tert-Butyltoluene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 192.8
Melting Pt: -62.5
Density: 0.8612 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008837
CAS Number: 98-51-1
EINECS: 202-675-9
UN: 2929
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4930 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR