2,6-Diisopropylaniline 90+%

Supplier: Thermo Scientific Chemicals

16941-1000 16941-5000
TS16941-5000EA 192 USD
TS16941-5000 TS16941-1000
2,6-Diisopropylaniline 90+%
2,6-Diisopropylaniline
Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.94 g/cm³
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00008887
CAS Number: 24544-04-5
EINECS: 246-305-4

Order Now

Specification Test Results

Appearance Colorless to yellow to orange to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5370 (20°C, 589 nm)
GC ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR