(4-Nitrophenyl)acetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitrobenzyl cyanide

16606-0250 16606-1000
TS16606-1000EA 81 USD
TS16606-1000 TS16606-0250
(4-Nitrophenyl)acetonitrile 98%
(4-Nitrophenyl)acetonitrile
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 195…197 °C (16hPa)
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007372
CAS Number: 555-21-5
EINECS: 209-085-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Cream to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 118 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR