3-Iodobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Iodobenzoic acid

16048-0100 16048-0500
TS16048-0100EA 36 USD
TS16048-0100 TS16048-0500
3-Iodobenzoic acid 98%
3-Iodobenzoic acid
Formula: C₇H₅IO₂
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9
EINECS: 210-555-2

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 185 to 189 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR