2-Bromobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Bromobenzoic Acid

16008-0250 16008-1000
TS16008-0250EA 41 USD
TS16008-0250 TS16008-1000
2-Bromobenzoic acid 97%
2-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 296
Melting Pt: 146…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 145 to 150 °C
HPLC ≥96.0 %
Fines ≤8.5 % (< 0.5 cm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR