(±)-1,2-Propanediol 99%, Extra Pure

Supplier: Thermo Scientific Chemicals

15872-0010 15872-0025 15872-0250
TS15872-0010EA 60 USD
TS15872-0010 TS15872-0025 TS15872-0250
(±)-1,2-Propanediol 99%, Extra Pure
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –60 °C
Density: 1.03 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
Water ≤0.2 % (Coulometric)
GC ≥98.5 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR