4-Chlorobenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15856-0050
15856-0250
15856-1000
15856-5000
TS15856-0050EA
29
USD
TS15856-0050
TS15856-0250
TS15856-1000
TS15856-5000
4-Chlorobenzenesulfonyl chloride 97%
4-Chlorobenzenesulphonyl chloride
Formula:
C₆H₄Cl₂O₂S Boiling Pt: 272 °C (1013 hPa) Melting Pt: 50…52 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007439 CAS Number: 98-60-2 EINECS: 202-685-3 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 50 to 55 °C |
Titration RCOCl | ≥96.0 % |
Titrable acid | ≤0.002 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...