2-Methylbenzylamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15563-0050
15563-0250
TS15563-0050EA
77
USD
TS15563-0050
TS15563-0250
2-Methylbenzylamine 96%
2-Methylbenzylamine
Formula:
C₈H₁₁N Boiling Pt: 196…197 °C (760 mmHg) Melting Pt: < 0 °C Density: 0.97 g/cm³ Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008112 CAS Number: 89-93-0 EINECS: 201-952-1 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5415 to 1.5435 (20°C, 589 nm) |
Titration with HClO4 | ≥95.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...