1-Dodecanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol
15545-0010
15545-0050
15545-0100
15545-5000
TS15545-0010EA
76
USD
TS15545-0010
TS15545-0050
TS15545-0100
TS15545-5000
1-Dodecanol 98%
1-Dodecanol
Formula:
H₃C(CH₂)₁₀CH₂OH Boiling Pt: 258…265 °C (760 mmHg) Melting Pt: 22…27 °C Density: 0.835 g/cm³ Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004753 CAS Number: 112-53-8 EINECS: 203-982-0 UN: 3077 ADR: 9,III Merck Index: 13,03439 |
Specification Test Results
Appearance | Colorless to white Liquid or crystalline solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...