α,α'-Dibromo-m-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

15505-0050 15505-0250
TS15505-0050EA 78 USD
TS15505-0050 TS15505-0250
α,α'-Dibromo-m-xylene 97%
α,α'-Dibromo-m-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 135…140 °C (20 mmHg)
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000178
CAS Number: 626-15-3
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 74 to 77 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR