2-Bromofluorene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15422-0250
15422-1000
TS15422-0250EA
126
USD
TS15422-0250
TS15422-1000
2-Bromofluorene 95%
2-Bromofluorene
Formula:
C₁₃H₉Br Boiling Pt: 185 °C (135 mmHg) Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001115 CAS Number: 1133-80-8 EINECS: 214-480-6 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 110 to 115 °C |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...