9-Anthrealdehyde 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde

15393-0250 15393-1000 15393-5000
TS15393-0250EA 65 USD
TS15393-0250 TS15393-1000 TS15393-5000
9-Anthrealdehyde 99%
9-Anthrealdehyde
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 369
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001254
CAS Number: 642-31-9
EINECS: 211-383-0

Order Now

Specification Test Results

Appearance Yellow to yellow-green to yellow-brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR