4-Chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Chlorobenzonitrile

15187-0250 15187-1000 15187-5000
TS15187-0250EA 51 USD
TS15187-0250 TS15187-1000 TS15187-5000
4-Chlorobenzonitrile 99%
4-Chlorobenzonitrile
Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 223
Melting Pt: 91…94 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4

Order Now

Specification Test Results

Appearance White to yellow Flakes and/or chunks
Infrared spectrum Conforms
Melting point 91 to 94 °C
Water ≤0.1 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR