Toluene-4-sulfonamide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
p-Toluenesulfonamide, 4-Methylbenzenesulfonamide, PTSA, 4-Toluenesulfonamide, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide
14769-0010
14769-0050
14769-2500
TS14769-0010EA
81
USD
TS14769-0010
TS14769-0050
TS14769-2500
Toluene-4-sulfonamide 99%
Toluene-4-sulphonamide
Formula:
C₇H₉NO₂S Boiling Pt: 221 °C (10 mmHg) Melting Pt: 136…140 °C Flash Pt: 202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011692 CAS Number: 70-55-3 EINECS: 200-741-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 140 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...