3-Aminophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Aminophenol3-Hydroxyaniline

14610-0010 14610-0050 14610-2500
TS14610-0010EA 204 USD
TS14610-0010 TS14610-0050 TS14610-2500
3-Aminophenol 99%
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 164 °C (15hPa)
Melting Pt: 121…124 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00007786
CAS Number: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to grey to light brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 120 to 124 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR