Methyl-4-hydroxybenzoate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl Parahydroxybenzoate
, Methyl 4-hydroxybenzoate
, 4-Hydroxybenzoic acid methyl ester
, Methylparaben
12696-0100
12696-0025
12696-5000
12696-1000
TS12696-5000EA
98.3
USD
TS12696-5000
TS12696-0025
TS12696-0100
TS12696-1000
Methyl-4-hydroxybenzoate 99%
Methyl 4-hydroxybenzoate
Formula:
HOC₆H₄CO₂CH₃ MW: 152.15 g/mol Boiling Pt: 298.6 °C (760 mmHg) Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002352 CAS Number: 99-76-3 EINECS: 202-785-7 Merck Index: 13,06129 |
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 125 to 128 °C |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...