1-Iodo-4-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

12238-0250 12238-1000
TS12238-0250EA 84 USD
TS12238-0250 TS12238-1000
1-Iodo-4-nitrobenzene 99%
1-Iodo-4-nitrobenzene
Formula: C₆H₄INO₂
Boiling Pt: 289 °C (772 mmHg)
Melting Pt: 171…173 °C
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7

Order Now

Specification Test Results

Appearance Light yellow to orange to brown Powder
Infrared spectrum Conforms
Melting point 172 to 177 °C
GC ≥98.5 %
UV at 270 nm A: ≤0.05
pH 8 to 9
Titration with HClO4 99.5 to 100.5 %
Chloride (Cl) ≤0.005 %
Iron (Fe) ≤0.0005 %
Lead (Pb) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR