3-Hydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

12098-0025 12098-1000 12098-5000
TS12098-0025EA 674 USD
TS12098-0025 TS12098-1000 TS12098-5000
3-Hydroxybenzoic acid 99%
3-Hydroxybenzoic acid
Formula: C₇H₆O₃
Melting Pt: 201…204 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 201 to 205 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR