Bis(4-fluorophenyl) sulfone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Fluor-4-((4-fluorphenyl)sulfonyl)benzol, 4-Fluorophenyl sulphone, 4,4'-Sulfonylbis(fluorobenzene), 4-Fluorophenyl sulfone, 4,4'-Difluoro diphenyl sulphone, 4,4'-Difluoro diphenyl sulfone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)

11962-0250 11962-1000
TS11962-0250EA 100 USD
TS11962-0250 TS11962-1000
Bis(4-fluorophenyl) sulfone 99%
Bis(4-fluorophenyl) sulphone
Formula: C₁₂H₈F₂O₂S
MW: 254.26 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000350
CAS Number: 383-29-9
EINECS: 206-847-4

Order Now

Specification Test Results

Appearance White to brown Powder or crystals
Infrared spectrum Conforms
Melting point 97 to 100 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR