1,4-Dimethoxybenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning

11541-0010 11541-1000 11541-5000
TS11541-0010EA 131 USD
TS11541-0010 TS11541-1000 TS11541-5000
1,4-Dimethoxybenzene 99+%
1,4-Dimethoxybenzene
Formula: C₈H₁₀O₂
Boiling Pt: 212.6 °C (1013 hPa)
Melting Pt: 54…58 °C
Density: 1.05 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00008401
CAS Number: 150-78-7
EINECS: 205-771-9

Order Now

Specification Test Results

Appearance White to off-white to grey Crystals of flakes
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR