4-Bromophenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid
10728-0050
10728-0250
TS10728-0050EA
81
USD
TS10728-0050
TS10728-0250
4-Bromophenylboronic acid 98%
4-Bromophenylboronic acid
Formula:
C₆H₆BBrO₂ Melting Pt: 287…292 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002104 CAS Number: 5467-74-3 EINECS: 226-779-9 |
Specification Test Results
Appearance | Off-white to light beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...