4-Bromobenzophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Bromophenyl)(phenyl)methanone, p-Bromobenzophenone
10672-0250
10672-1000
TS10672-0250EA
43
USD
TS10672-0250
TS10672-1000
4-Bromobenzophenone 97%
4-Bromobenzophenone
Formula:
C₁₃H₉BrO Boiling Pt: 350 °C (1013 hPa) Melting Pt: 78…82 °C |
MDL Number:
MFCD00000103 CAS Number: 90-90-4 EINECS: 202-024-9 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 78 to 82 °C |
GC | ≥98.5 % |
Color scale | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...