2-Phenylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid
10627-0250
10627-1000
TS10627-0250EA
68
USD
TS10627-0250
TS10627-1000
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol Boiling Pt: 199 °C (10 mmHg) Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002463 CAS Number: 947-84-2 EINECS: 213-432-1 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 110 to 114 °C |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...