Pentafluoro(phenoxy)cyclotriphosphazene ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine

P2286-1G P2286-5G
TCP2286-1GEA 139 USD
TCP2286-1G TCP2286-5G
Pentafluoro(phenoxy)cyclotriphosphazene ≥98.0% (by GC)
Pentafluoro(phenoxy)cyclotriphosphazene
Formula: C₆H₅F₅N₃OP₃
Boiling Pt: 46°C
Storage Temperature: Refrigerator
CAS Number: 33027-68-8
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR