1,3,6-Hexanetricarbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Hexane-1,3,6-tricarbonitrile

H1504-25G H1504-5G
TCH1504-25GEA 738.52 USD
TCH1504-25G TCH1504-5G
1,3,6-Hexanetricarbonitrile ≥98.0% (by GC)
1,3,6-Hexanetricarbonitrile
Formula: C₉H₁₁N₃
MW: 161.21 g/mol
Boiling Pt: 200°C
Flash Pt: 85°C
MDL Number: MFCD00129792
CAS Number: 1772-25-4
EINECS: 217-199-7
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR