United Scientific™ Intermediate Spectrometer with 30 mm Optics
Supplier: United Scientific Supplies
SPECT02-P SPECT03
470339-896EA 172.29 USD
470339-896 470340-220
United Scientific™ Intermediate Spectrometer with 30 mm Optics
Spectrophotometers and Photometers UV/Visible Spectrophotometers
High quality spectroscope for observing spectral lines.
  • Easy-to-use classroom spectrometer
  • 30 mm diameter optics give brighter line images and make precise measurements easier
This instrument is an enhanced version of United Scientific's popular Intermediate Spectrometer (SPECT02.) Its larger 30 mm diameter optics give brighter line images and make precise measurements easier. The optical focusing system has also been refined and the instrument retains the same premium features as the earlier version, including the built-in illuminated autocollimator for fast set up, 17.8 cm diameter divided circle, double-ended vernier reading to 1' resolution, and 7 cm diameter rotatable and height-adjusting prism table to accommodate various accessories.
Delivery information: Supplied with a 300 lines/mm diffraction grating and grating holder, high quality prism, illuminated magnifier, auto-collimation mirror with a power supply for 110 VAC, and an instruction manual