Innovating Science® Urinalysis using Simulated Urine
Supplier: Ward's Science
IS1513 IS3008
470330-226EA 31.5 USD
470330-226 470343-478
Innovating Science® Urinalysis using Simulated Urine
Educational Classroom Kits and Activities
Learn about urinalysis, one of the oldest medical diagnostic tests performed.
  • Students will use simulated urine to test factors like pH, color, clarity, and test for the presence of factors such as proteins, glucose, and calcium
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Urinalysis, one of the oldest medical diagnostic tests performed, is to this day still one of the most common. In this activity, students will use simulated urine to avoid the unpleasantness of using the real thing while still performing actual tests used on real urine samples. Students will examine the simulated urine for factors such as pH, color, clarity, as well as test for the presence or absence of proteins, glucose, and calcium.
The students will then examine the samples microscopically to determine if crystals may be present in any of the samples. Kit contains enough materials for 15 groups of students. Teacher's Manual and Student Study Guide copymasters are included. Needed but not supplied are a hot water bath, glass test tubes, and compound microscopes (4X/10X/40X).
Delivery information: Instructor's Manual and SDS included.