Measurement Kit
Supplier: United Scientific Supplies
MEAKIT
470340-210EA 77.75 USD
470340-210
Measurement Kit
Educational Materials Math Educational Materials Measurement Learning Activities
Basic variety of measuring tools with guides and activities.
  • Variety of quality tools and objects for teaching basics of measurement
  • Shipped in a reusable storage box
  • Includes guide with 15 activities
This kit includes a variety of tools and objects that are ideal for teaching the basics of measurement.
Kit contents: Depth gauge; Micrometer; Plastic protractor; Plastic ruler (12"); Tape measure; Vernier caliper, brass; Wire gauge with conversion table; Aluminum square; Aluminum wire; Graduated cylinder; Plastic cylinder; Plastic vial.