2-(Hydroxymethyl)indole 96%

Supplier: Thermo Scientific Chemicals
Danger

45981-0010 45981-0050
TS45981-0010EA 100 USD
TS45981-0010 TS45981-0050
2-(Hydroxymethyl)indole 96%
2-(Hydroxymethyl)indole
Formula: C₉H₉NO
Melting Pt: 72…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00030195
CAS Number: 24621-70-3

Order Now

Specification Test Results

Appearance White to light yellow to tan Solid
Infrared spectrum Conforms
Melting point 72 to 78 °C
Water ≤1 %
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR