4-Bromo-1-butanol 85+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Bromo-4-hydroxybutane

39001-0010 39001-0050 39001-0250
TS39001-0010EA 73 USD
TS39001-0010 TS39001-0050 TS39001-0250
4-Bromo-1-butanol 85+%
4-Bromo-1-butanol
Formula: BrH₂C(CH₂)₂CH₂OH
MW: 153.02
Boiling Pt: 56…58 °C (2 mmHg)
Flash Pt: 7 °C
Storage Temperature: Freezer
MDL Number: MFCD00015387
CAS Number: 33036-62-3
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to purple to brown Liquid
Infrared spectrum Conforms
GC ≥85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR