5-Allyl-3-methoxysalicylaldehyde 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Allyl-2-hydroxy-3-methoxybenzaldehyde
37936-0100
37936-0250
TS37936-0100EA
88
USD
TS37936-0100
TS37936-0250
5-Allyl-3-methoxysalicylaldehyde 98%
5-Allyl-3-methoxysalicylaldehyde
Formula:
C₁₁H₁₂O₃ Melting Pt: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD06408022 CAS Number: 22934-51-6 |
Specification Test Results
Appearance | Light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 48 to 52 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...