(4-Nitrophenyl)-α-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Nitrophenyl α-D-glucopyranoside(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol4-Nitrophenyl a-D-glucopyranoside

33715-0050 33715-0010 33715-0250
TS33715-0010EA 66.3 USD
TS33715-0010 TS33715-0250 TS33715-0050
(4-Nitrophenyl)-α-D-glucopyranoside 99%
(4-Nitrophenyl) α-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 204…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064088
CAS Number: 3767-28-0
EINECS: 223-189-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (2% in methanol) Clear to slightly hazy colorless to green-yellow solution
HPLC ≥98.5 %
Specific optical rotation +210° to +220° (20°C, 589 nm) (c=1, water)
Impurity ≤0.1 % (p-Nitrophenol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR