(±)-3-Methyl-1-pentanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-3-Methylpentan-1-ol3-Methyl-1-pentanol3-Methylpentan-1-ol

33532-0050 33532-0250 33532-1000
TS33532-0050EA 29.9 USD
TS33532-0050 TS33532-1000 TS33532-0250
(±)-3-Methyl-1-pentanol 99+%
(±)-3-Methyl-1-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 150…154 °C
Melting Pt: –70 °C
Density: 0.823 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002937
CAS Number: 589-35-5
EINECS: 209-644-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR