(R)-(-)-1-Methoxy-2-propanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1-Methoxy-2-propanol, (R)-(-)-Propylene glycol 1-Monomethyl ether

33178-0010 33178-0100
TS33178-0010EA 46 USD
TS33178-0010 TS33178-0100
(R)-(-)-1-Methoxy-2-propanol 98+%
(R)-(-)-1-Methoxy-2-propanol
Formula: C₄H₁₀O₂
Melting Pt: –97 °C
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD01632587
CAS Number: 4984-22-9
UN: 3092
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20 °C, 589 nm)
GC ≥98.0 %
Specific optical rotation -20° to -24° (24°C, 589 nm) (c=10, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR