Octyl acetate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid octyl ester

31063-0050 31063-1000 31063-5000
TS31063-0050EA 23 USD
TS31063-0050 TS31063-1000 TS31063-5000
Octyl acetate 99+%
Octyl acetate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 211 °C (760 mmHg)
Melting Pt: -38.5
Density: 0.868 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00009562
CAS Number: 112-14-1
EINECS: 203-939-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4175 to 1.4195 (20 °C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR