(±)-Camphene 75% remainder mainly alpha-fenchene

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-3,3-Dimethyl-2-methylennorcamphaneCamphene(±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane

29557-0010 29557-0025
TS29557-0010EA 112 USD
TS29557-0010 TS29557-0025
(±)-Camphene 75% remainder mainly alpha-fenchene
(±)-Camphene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 159…160 °C (760 mmHg)
Melting Pt: 42 °C
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00066603
CAS Number: 79-92-5
EINECS: 201-234-8
UN: 1325
ADR: 4.1,II
Merck Index: 12,01777

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
GC ≥70 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR