Sesame oil

Supplier: Thermo Scientific Chemicals

24100-0010 24100-2500
TS24100-0010EA 167 USD
TS24100-0010 TS24100-2500
Sesame oil
Sesame oil
Melting Pt: –5 °C
Density: 0.919 g/cm³
Flash Pt: 255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00132243
CAS Number: 8008-74-0

Order Now

Specification Test Results

Appearance Clear yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4760 (20 °C, 589 nm)
Density (20°C) 0.915 to 0.923
Acid value ≤0.6 mg KOH/g
Peroxide value ≤10 meq/kg
Unsaponifiable matter ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR