(±)-α-Terpineol 95% (α-isomer) (mixture of alpha- and gamma-terpineol)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
, Terpineol
20374-0500
20374-2500
TS20374-2500EA
100
USD
TS20374-2500
TS20374-0500
(±)-α-Terpineol 95% (α-isomer) (mixture of alpha- and gamma-terpineol)
(±)-α-Terpineol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Melting Pt: 18 °C Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001557 CAS Number: 98-55-5 EINECS: 232-268-1 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4800 to 1.4850 (20 °C, 589 nm) |
Density | (20°C) 0.930 to 0.938 g/ml |
GC | ≥94 % (alpha + gamma) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...