Paraffin, liquid, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Wax Paraffin
, Paraffin wax
17140-2500
17140-0025
17140-0010
TS17140-0025EA
150
USD
TS17140-0025
TS17140-2500
TS17140-0010
Paraffin, liquid, pure
Paraffin
Boiling Pt:
300 °C (760 mmHg) Melting Pt: –24 °C Density: 0.88 g/cm³ Flash Pt: 218 °C Storage Temperature: Ambient |
CAS Number:
8002-74-2 |
Specification Test Results
Appearance | Clear colorless Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4700 to 1.4800 (20 °C, 589 nm) |
Density | (20°C) 0.827 to 0.890 g/ml |
Viscosity | 63 to 80 mm²/s (40°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...