1-Acetyladamantane 99%

Supplier: Thermo Scientific Chemicals

16474-0010 16474-0050 16474-0250
TS16474-0010EA 32 USD
TS16474-0010 TS16474-0050 TS16474-0250
1-Acetyladamantane 99%
1-Acetyladamantane
Formula: C₁₂H₁₈O
Melting Pt: 53…55 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00074739
CAS Number: 1660-04-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 55 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR