Pregnenolone 99%

Supplier: Thermo Scientific Chemicals

16442-0050 16442-0250 16442-1000
TS16442-0050EA 44 USD
TS16442-0050 TS16442-0250 TS16442-1000
Pregnenolone 99%
Pregnenolone
Formula: C₂₁H₃₂O₂
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00003628
CAS Number: 145-13-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
GC ≥98.5 %
Specific optical rotation +27° to +30° (20°C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR