α-Pinene 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with α-tocopherol
16436-0050
16436-2500
16436-0010
TS16436-0010EA
310
USD
TS16436-0010
TS16436-0050
TS16436-2500
α-Pinene 97% stabilized
α-Pinene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 154…157 °C (760 mmHg) Melting Pt: –55 °C Density: 0.85 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001339 CAS Number: 80-56-8 EINECS: 219-445-9 UN: 2368 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4640 to 1.4680 (20 °C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | 0.8 % Alpha-tocopherol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...