Diethyl oxalate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Oxalic acid diethyl ester

15918-0025 15918-5000 15918-0100 15918-0250 15918-0010
TS15918-0010EA 61 USD
TS15918-0010 TS15918-0250 TS15918-5000 TS15918-0025 TS15918-0100
Diethyl oxalate 99%
Diethyl oxalate
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185 °C (760 mmHg)
Melting Pt: –41 °C
Density: 1.07 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4085 to 1.4105 (20 °C, 589 nm)
Water ≤0.05 %
Density (20°C) 1.078 to 1.082 kg/l
GC ≥98.5 %
Color scale ≤15 APHA
Acidity ≤0.01 % (as oxalic acid)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR