Dimethyl succinate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester

15073-1000 15073-5000
TS15073-1000EA 48 USD
TS15073-1000 TS15073-5000
Dimethyl succinate 99%
Dimethyl succinate
Formula: C₆H₁₀O₄
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 18…19 °C
Density: 1.11 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008466
CAS Number: 106-65-0
EINECS: 203-419-9
Merck Index: 12,09038

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4210 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤25 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR