Sulfolane 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetramethylene sulfone, Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene-1,1-dioxide, Tetrahydrothiophene 1,1-dioxide

13843-0010 13843-0025 13843-0100 13843-2500
TS13843-0010EA 155 USD
TS13843-0010 TS13843-0025 TS13843-0100 TS13843-2500
Sulfolane 99+%
Sulpholane
Formula: C₄H₈O₂S
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 27.4…27.8 °C
Density: 1.26 g/cm³
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4820 to 1.4850 (20 °C, 589 nm)
Water ≤0.3 %
Sulfated ash ≤0.1 %
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR