Pyridine-2-carbaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13182-0250
13182-1000
13182-5000
TS13182-0250EA
38
USD
TS13182-0250
TS13182-1000
TS13182-5000
Pyridine-2-carbaldehyde 99%
Pyridine-2-carbaldehyde
Formula:
C₆H₅NO Boiling Pt: 180…182 °C (761 mmHg) Density: 1.121 g/cm³ Flash Pt: 73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006290 CAS Number: 1121-60-4 EINECS: 214-333-6 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5343 to 1.5373 (20 °C, 589 nm) |
GC | ≥99.5 % |
UV | at 220 nm A: ≤0.7 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Carbonyl compounds | ≤0.03 % (as C2H5CHO) |
Isopropyl alcohol | ≤0.05 % |
Titrable acid | 0.0004 meq/g |
Ethanol | ≤0.01 % |
Methanol | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...