Oleyl alcohol ca. 60%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

12953-0010 12953-0025
TS12953-0010EA 118 USD
TS12953-0010 TS12953-0025
Oleyl alcohol ca. 60%, Technical Grade
Oleyl alcohol
Formula: C₁₈H₃₆O
Boiling Pt: 207 °C (13 mmHg)
Melting Pt: 5 °C
Density: 0.84 g/cm³
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002993
CAS Number: 143-28-2
EINECS: 205-597-3
Merck Index: 12,06968

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4640 (20 °C, 589 nm)
GC 60 % Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR