Caprylaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Octanal, Octylaldehyde, Octaldehyde

12948-1000 12948-5000
TS12948-1000EA 44 USD
TS12948-1000 TS12948-5000
Caprylaldehyde 99%
Caprylaldehyde
Formula: C₈H₁₆O
Boiling Pt: ∼171 °C (1013 hPa)
Melting Pt: 12…15 °C
Density: 0.821 g/cm³
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007029
CAS Number: 124-13-0
EINECS: 204-683-8
UN: 1191
ADR: 3,III
Merck Index: 13,01772

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4200 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR