(±)-Linalool 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Dimethylocta-1,6-dien-3-ol(±)-3,7-Dimethyl-1,6-octadien-3-ol

12515-5000 12515-0025 12515-1000
TS12515-5000EA 137 USD
TS12515-5000 TS12515-0025 TS12515-1000
(±)-Linalool 97%
(±)-Linalool
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 199 °C (760 mmHg)
Density: 0.861 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008906
CAS Number: 78-70-6
EINECS: 201-134-4
Merck Index: 13,05517

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20 °C, 589 nm)
GC ≥96.0 %
Specific gravity (20/4°C) 0.859 to 0.864
Specific optical rotation -2° to +2° (20°C, 589 nm) (neat)
Substance darkened by H2SO4 ≤10 APHA
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.2 ppm
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.001 %
Bromine number ≤0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR